Hoechst 33342 alkyne
Lieferant: AAT BIOQUEST
17673
AATB17673EA
402
EUR
AATB17673
Hoechst 33342 alkyne
Hoechst 33342 alkyne
Hoechst 33342 alkyne is an excellent building block for developing site-specific nucleic acid probes via the well-known click chemistry, i.e., Copper-Catalyzed Azide–Alkyne Cycloaddition (CuAAC).
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only