Hoechst 33342 azide
Lieferant: AAT BIOQUEST
17674
AATB17674EA
402
EUR
AATB17674
Hoechst 33342 azide
Hoechst 33342 azide
Hoechst 33342 azide is an excellent building block for developing site-specific nucleic acid probes via the well-known click chemistry, i.e., Copper-Catalyzed Azide–Alkyne Cycloaddition (CuAAC).
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only