Gelite™ Safe DNA Gel Stain *10000X Water Solution*
Lieferant: AAT BIOQUEST
17699
AATB17699EA 22.2 EUR
AATB17699
Gelite™ Safe DNA Gel Stain *10000X Water Solution*
Färbemittel Elektrophorese-Färbemittel
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
  • Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Bitte beachten For research use only.