Gelite™ Safe DNA Gel Stain *GelRed Alternative, 10000X in DMSO*
Lieferant: AAT BIOQUEST
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
- Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Bitte beachten For research use only.