Gelite™ 50 bp DNA Ladder
Lieferant: AAT BIOQUEST
17750
AATB17750EA 133 EUR
AATB17750
Gelite™ 50 bp DNA Ladder
Elektrophorese-Reagenzien Molekulargewichtsmarker
The Gelite™ 50 bp DNA Ladder is ideal for sizing and approximating the mass of double-stranded DNA within the 50 to 1500 bp range on agarose or polyacrylamide gels.
  • Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.