Gelite™ 50 bp DNA Ladder
Lieferant: AAT BIOQUEST
17750
AATB17750EA
133
EUR
AATB17750
Gelite™ 50 bp DNA Ladder
Elektrophorese-Reagenzien
Molekulargewichtsmarker
The Gelite™ 50 bp DNA Ladder is ideal for sizing and approximating the mass of double-stranded DNA within the 50 to 1500 bp range on agarose or polyacrylamide gels.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.