XFD350 acid fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1786
AATB1786EA
439
EUR
AATB1786
XFD350 acid fluoreszierende Probe
XFD350 acid
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
313,28 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...