XFD488 acid fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1790
AATB1790EA 460 EUR
AATB1790
XFD488 acid fluoreszierende Probe
XFD488 acid

XFD488 has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 648,49 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR