XFD546 acid fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1791
AATB1791EA 439 EUR
AATB1791
XFD546 acid fluoreszierende Probe
XFD546 acid

XFD546 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 546 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 1264,91 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR