XFD514 acid
Lieferant: AAT BIOQUEST
1793
AATB1793EA 429 EUR
AATB1793
XFD514 acid
XFD514 acid
XFD514 is manufactured by AAT Bioquest, and it has the same chemical structure to Alexa Fluor® 514 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only