XFD700 acid
Lieferant: AAT BIOQUEST
1799
AATB1799EA
619
EUR
AATB1799
XFD700 acid
XFD700 acid
XFD700 is the same molecule as the Alexa Fluor® 700 (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house