ICG-Sulpho-OSu ≥80%
Lieferant: AAT BIOQUEST
180
AATB180EA 460 EUR
AATB180
ICG-Sulpho-OSu ≥80%
ICG-Sulpho-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
  • Dependable in vivo imaging applications
  • Infrared absorption wavelength
  • Ideal for bioconjugation
Bitte beachten For research use only.
CAS-Nummer: 190714-28-4