XFD405 NHS ester [equivalent to Alexa Fluor™ 405 NHS ester]
Lieferant: AAT BIOQUEST
1804
AATB1804EA
341
EUR
AATB1804
XFD405 NHS ester [equivalent to Alexa Fluor™ 405 NHS ester]
XFD405 NHS ester
XFD405 NHS Ester (Succinimidyl Ester) is the same molecule as Alexa Fluor® 405 NHS Ester (#A30000 from ThermoFisher, Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only