3-ICG-acyl-1,3-thiazolidine-2-thione (ICG-ATT) ≥90%
Lieferant: AAT BIOQUEST
181
AATB181EA 614 EUR
AATB181
3-ICG-acyl-1,3-thiazolidine-2-thione (ICG-ATT) ≥90%
3-ICG-acyl-1,3-thiazolidine-2-thione (ICG-ATT)
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
  • Dependable invivo imaging applications
  • Infrared absorption wavelength
  • Ideal for bioconjugation
Bitte beachten For research use only.