XFD532 NHS Ester fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1819
AATB1819EA
307
EUR
AATB1819
XFD532 NHS Ester fluoreszierende Probe
XFD532 NHS Ester
XFD532 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 532 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
723,77 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...