ICG-OSu ≥90%

Lieferant: AAT BIOQUEST

182
AATB182EA 230 EUR
AATB182
ICG-OSu ≥90%
ICG-OSu

Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.


  • Dependable in vivo imaging applications
  • Infrared absorption wavelength
  • Ideal for bioconjugation


Bitte beachten For research use only.

Molecular Weight: 828,03 g/mol
CAS-Nummer: 1622335-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR