ICG-OSu ≥97%, ultrarein
Lieferant: AAT Bioquest
182-P97
AATB182-P97EA 470 EUR
AATB182-P97
ICG-OSu ≥97%, ultrarein
ICG-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics for determining cardiac output, hepatic function, and liver blood flow, as well as ophthalmic angiography.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only
CAS-Nummer: 1622335-40-3