XFD546 NHS Ester fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1820
AATB1820EA
292
EUR
AATB1820
XFD546 NHS Ester fluoreszierende Probe
XFD546 NHS Ester
XFD546 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 546 (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
1159,6 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...