XFD568 NHS Ester fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1823
AATB1823EA 292 EUR
AATB1823
XFD568 NHS Ester fluoreszierende Probe
XFD568 NHS Ester

XFD568 has the same chemical structure of Alexa Fluor® 568 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 791,8 g/mol
CAS-Nummer: 247145-38-6
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR