ICG-Sulpho-EG4-OSu ≥90%
Lieferant: AAT BIOQUEST
183
AATB183EA
537
EUR
AATB183
ICG-Sulpho-EG4-OSu ≥90%
ICG-Sulpho-EG4-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
- Dependable in vivo imaging applications
- Infrared absorption wavelength
- Ideal for bioconjugation
- Improved water solubility
Bitte beachten For research use only.