XFD594 NHS Ester fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1830
AATB1830EA 307 EUR
AATB1830
XFD594 NHS Ester fluoreszierende Probe
XFD594 NHS Ester

XFD594 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 594 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 819,85 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR