ICG-Sulpho-EG8-OSu ≥90%
Lieferant: AAT BIOQUEST
184
AATB184EA
537
EUR
AATB184
ICG-Sulpho-EG8-OSu ≥90%
ICG-Sulpho-EG8-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
- Dependable in vivo imaging applications
- Infrared absorption wavelength
- Ideal for bioconjugation
- Improved water solubility
Bitte beachten For research use only.