ICG-Sulpho-EG8-OSu ≥90%
Lieferant: AAT BIOQUEST
184
AATB184EA 537 EUR
AATB184
ICG-Sulpho-EG8-OSu ≥90%
ICG-Sulpho-EG8-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
  • Dependable in vivo imaging applications
  • Infrared absorption wavelength
  • Ideal for bioconjugation
  • Improved water solubility
Bitte beachten For research use only.