ICG-PEG12-OSu ≥90%
Lieferant: AAT BIOQUEST
185
AATB185EA 614 EUR
AATB185
ICG-PEG12-OSu ≥90%
ICG-PEG12-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
  • Dependable invivo imaging applications
  • Infrared absorption wavelength
  • Ideal for bioconjugation
Bitte beachten For research use only.