ICG-PEG12-OSu ≥90%
Lieferant: AAT BIOQUEST
185
AATB185EA
614
EUR
AATB185
ICG-PEG12-OSu ≥90%
ICG-PEG12-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
- Dependable invivo imaging applications
- Infrared absorption wavelength
- Ideal for bioconjugation
Bitte beachten For research use only.