ICG Xtra-OSu ≥90%
Lieferant: AAT BIOQUEST
186
AATB186EA
307
EUR
AATB186
ICG Xtra-OSu ≥90%
ICG Xtra-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
- Dependable in vivo imaging applications
- Infrared absorption wavelength
- Ideal for bioconjugation
Bitte beachten For research use only.