XFD350 C5 Maleimide fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1870
AATB1870EA
307
EUR
AATB1870
XFD350 C5 Maleimide fluoreszierende Probe
XFD350 C5 Maleimide
XFD350 C5 maleimide is the same molecule to the Alexa Fluor® 350 C5 maleimide (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
578,68 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...