XFD532 C5 Maleimide fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1885
AATB1885EA
307
EUR
AATB1885
XFD532 C5 Maleimide fluoreszierende Probe
XFD532 C5 Maleimide
XFD532 C5 Maleimide is the same molecule to the Alexa Fluor® C5 Maleimide (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
812,88 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...