ICG acid ≥95%
Lieferant: AAT BIOQUEST
189
AATB189EA
307
EUR
AATB189
ICG acid ≥95%
ICG acid
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
- Dependable in vivo imaging applications
- Infrared absorption wavelength
- Ideal for bioconjugation
Bitte beachten For research use only.