XFD594 C5 Maleimide fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1891
AATB1891EA 307 EUR
AATB1891
XFD594 C5 Maleimide fluoreszierende Probe
XFD594 C5 Maleimide

XFD594 C5 Maleimide is the same molecule to Alexa Fluor® 594 C5 Maleimide (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal

Molecular Weight: 908,97 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd8a46bad00f9438fd7d8250d73b81cce245b17e&vid=1eb7c136d392b3ae45c6d1a0b717d5b1832853c9&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR