XFD647 C2 maleimide [Same Structure to Alexa Fluor™ 647 C2 maleimide]
Lieferant: AAT BIOQUEST
1895
AATB1895EA 309 EUR
AATB1895
XFD647 C2 maleimide [Same Structure to Alexa Fluor™ 647 C2 maleimide]
XFD647 C2 maleimide
XFD647 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 647 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only