XFD647 azide
Lieferant: AAT BIOQUEST
1896
AATB1896EA 277 EUR
AATB1896
XFD647 azide
XFD647 azide
XFD647 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 647 (Alexa Fluor® is the trademark of ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only