Cy3NS acid ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Cyanine3 carboxylic acid chloride, 2-(3-(1-(5-Carboxypentyl)-3,3-dimethylindolin-2-ylidene)prop-1-en-1-yl)-1,3,3-trimethyl-3H-indol-1-ium chloride

190
AATB190EA 614 EUR
AATB190
Cy3NS acid ≥95%
Cy3NS acid

Cyanines are used in a wide variety of biological applications including comparative genomic hybridization and in gene chips.

Molecular Weight: 456,62 g/mol
MDL: MFCD28385499
CAS-Nummer: 1032678-01-5
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR