XFD488 Hydroxylamine fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1900
AATB1900EA
460
EUR
AATB1900
XFD488 Hydroxylamine fluoreszierende Probe
XFD488 Hydroxylamine
XFD488 Hydroxylamine is the same molecule to Alexa Fluor® 488 Hydroxylamine with higher purity (Alexa Fluor® is the trademark of ThermoFisher).
- pH-insensitive
- High photostability
- Brighter signal
Molecular Weight:
894,07 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...