ReadiLink™ xtra Rapid XFD594 Antibody Labelling Kit *BSA-Compatible, XFD594 Same Structure to Alexa Fluor™ 594* (2 Labellings)
Lieferant: AAT BIOQUEST
1982
AATB1982EA 292 EUR
AATB1982
ReadiLink™ xtra Rapid XFD594 Antibody Labelling Kit *BSA-Compatible, XFD594 Same Structure to Alexa Fluor™ 594* (2 Labellings)
Proteinkonjugate und Markierungen
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labelling protocol
  • No column purification required
Bitte beachten For research use only.