ReadiLink™ xtra Rapid XFD647 Antibody Labelling Kit *BSA-Compatible, XFD647 Same Structure to Alexa Fluor™ 647* (2 Labelings)
Lieferant: AAT BIOQUEST
1985
AATB1985EA 292 EUR
AATB1985
ReadiLink™ xtra Rapid XFD647 Antibody Labelling Kit *BSA-Compatible, XFD647 Same Structure to Alexa Fluor™ 647* (2 Labelings)
Proteinkonjugate und Markierungen
XFD647 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 647 (Alexa Fluor® is the trademark of ThermoFisher).
  • Contains all essential components
  • Minimal hands on time
  • Ready to use
  • Robust and simple antibody labeling protocol
  • No column purification required
Bitte beachten For research use only.