Uranin ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-Hydroxy-9-(2-carboxyphenyl)-(3H)-xanthen-3-on Dinatriumsalz, 6-Hydroxy-9-(2-carboxyphenyl)- (3H)-xanthen-3-on Dinatriumsalz, Fluoreszein Dinatriumsalz, Fluorescein Natriumsalz, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)-benzoesäure Dinatriumsalz, Dinatrium-2-(3-oxo-6-oxidoxanthen-9-yl)benzoat, 6-Hydroxy-9-(2-carboxyphenyl)-(3H)-3-xanthenon Dinatriumsalz, Fluorescein-Natrium, Resorcinphthalein Dinatriumsalz, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoesäure Dinatriumsalz, Fluorescein Dinatriumsalz, 3′,6′-Dihydroxyspiro[2-benzofuran-3,9′-xanthen]-1-on Dinatriumsalz, 9-(2-Carboxyphenyl)-6-hydroxy-(3H)-3-xanthenon Dinatriumsalz

2
AATB2EA 120 EUR
AATB2
Uranin ≥95%
Uranin

Fluorescein, disodium salt is water-soluble version of fluorescein (see product #1).

Formel: C₂₀H₁₀Na₂O₅
Molecular Weight: 376,28 g/mol
MDL: MFCD00167039
CAS-Nummer: 518-47-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR