5-(4,6-Dichlorotriazinyl)aminofluorescein für die Molekularbiologie

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 5-DTAF

200
AATB200EA 153 EUR
AATB200
5-(4,6-Dichlorotriazinyl)aminofluorescein für die Molekularbiologie
5-(4,6-Dichlorotriazinyl)aminofluorescein

The 5-isomer of fluorescein dichlorotriazine (5-DTAF) is highly reactive with proteins.


  • Useful for multiple applications

Formel: C₂₃H₁₂Cl₂N₄O₅
Molecular Weight: 495.28 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 51306-35-5
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR