FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit)
Lieferant: AAT BIOQUEST
20001
AATB20001EA 152 EUR
AATB20001
FluoroQuest™ Anti-Fading Kit I *Optimized for Slide Imaging* (1 Kit)
Färbemittel Mikroskopie- und Färbereagenzien
When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.
  • Preserves fluorescence
  • Prevents photobleaching
  • Ready to use
  • Contains all necessary components
Bitte beachten For research use only.