FluoroQuest™ Anti-Fading Kit II *Optimized for Plate Imaging* (1 Kits)
Lieferant: AAT BIOQUEST
20003
AATB20003EA
152
EUR
AATB20003
FluoroQuest™ Anti-Fading Kit II *Optimized for Plate Imaging* (1 Kits)
Färbemittel
Mikroskopie- und Färbereagenzien
When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.
- Preserves fluorescence
- Prevents photobleaching
- Ready to use
- Contains all necessary components
Bitte beachten For research use only.