FluoroQuest™ Anti-Fading Kit II *Optimized for Plate Imaging* (1 Kits)
Lieferant: AAT BIOQUEST
20003
AATB20003EA 152 EUR
AATB20003
FluoroQuest™ Anti-Fading Kit II *Optimized for Plate Imaging* (1 Kits)
Färbemittel Mikroskopie- und Färbereagenzien
When exposed to excitation light, fluorescence intensity of dyes decreases due to their photooxidation or other photoreactions.
  • Preserves fluorescence
  • Prevents photobleaching
  • Ready to use
  • Contains all necessary components
Bitte beachten For research use only.