FluoroQuest™ Mounting Medium with DAPI (50 ml)
Lieferant: AAT BIOQUEST
20004
AATB20004EA 152 EUR
AATB20004
FluoroQuest™ Mounting Medium with DAPI (50 ml)
Färbemittel Mikroskopie- und Färbereagenzien
FluoroQuest™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Contains DNA/RNA counterstain DAPI
  • Improves and sustains fluorescence
  • Stable for extended storage