FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml)
Lieferant: AAT BIOQUEST
20005
AATB20005EA
292
EUR
AATB20005
FluoroQuest™ Anti-Fading Mounting Medium with DAPI (20 ml)
Färbemittel
Mikroskopie- und Färbereagenzien
FluoroQuest™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
- Preserves fluorescence
- Prevents photobleaching
- Contains nuclear counterstain for convenience
Bitte beachten For research use only.