FluoroQuest™ PLUS Antifade Mounting Medium
Lieferant: AAT BIOQUEST
FluoroQuest™ PLUS Antifade mounting medium, the latest breakthrough in non-setting media technology, provides unparalleled protection against fading throughout the visible spectrum, including far-red wavelengths.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.