FluoroQuest™ PLUS Antifade Mounting Medium
Lieferant: AAT BIOQUEST
20008
AATB20008EA
180
EUR
AATB20008
FluoroQuest™ PLUS Antifade Mounting Medium
Färbemittel
Mikroskopie- und Färbereagenzien
FluoroQuest™ PLUS Antifade mounting medium, the latest breakthrough in non-setting media technology, provides unparalleled protection against fading throughout the visible spectrum, including far-red wavelengths.
- Stable for 12 months
Quality tested in-house.
Bitte beachten For research use only.