4-{[4-(Dimethylamino)phenyl]diazenyl}benzoesäure ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 4-Dimethylaminoazobenzene-4-carboxylic Acid, DABCYL acid, Para Methyl Red, p-Methyl Red

2001
AATB2001EA 230 EUR
AATB2001
4-{[4-(Dimethylamino)phenyl]diazenyl}benzoesäure ≥95%
4-{[4-(Dimethylamino)phenyl]diazenyl}benzoesäure

DABCYL acid is one of the most popular acceptors for developing FRET-based nucleic acid probes suh as Molecular Beacon-based PCR probes.

Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269,30 g/mol
MDL: MFCD00058988
CAS-Nummer: 6268-49-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR