Formaldehyd solution_SC 4%, ReadiUse™ Fixative
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Methanal, Formalin, Formol, Methylaldehyd
20010
AATB20010EA
120
EUR
AATB20010
Formaldehyd solution_SC 4%, ReadiUse™ Fixative
Formaldehyd
All our ReadiUse™ reagents require minimal hands-on time.
- Non-reactive
- Long-term stability
- Minimal hands-on time
- Ready to use
Bitte beachten For research use only.
Formel:
CH₂O Molecular Weight: 30.03 g/mol Siedepunkt: -19.3 Schmelzpunkt: -92 |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 ADR: 5 II Merck Index: 13,04259 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...