Pluronic® F-127 Cell Culture Tested
Lieferant: AAT BIOQUEST
20050
AATB20050EA 120 EUR
AATB20050
Pluronic® F-127 Cell Culture Tested
Reinigungsmittel Waschmittel und Oberflächenreiniger
Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.
  • Non-toxic to cells
It is soluble at low temperature, but gels at room temperature.