Pluronic® F-127 20% in DMSO

Lieferant: AAT BIOQUEST
Achtung

20052
AATB20052EA 120 EUR
AATB20052
Pluronic® F-127 20% in DMSO
Poloxamer 407 (Pluronic® F-127)

Pluronic® F-127 is a nonionic surfactant that is 100% active and relatively non-toxic to cells, and frequently used with dye AM esters to improve their water solubility.


  • Extremely common surfectant
  • Multiple applications
  • DMSO solution


Bitte beachten For research use only.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR