DABCYL C2 amine FRET-Akzeptor-Vorläufer

Lieferant: AAT BIOQUEST
Achtung

2006
AATB2006EA 307 EUR
AATB2006
DABCYL C2 amine FRET-Akzeptor-Vorläufer
DABCYL C2 amine

DABCYL C2 amine is the carbonyl-reactive form of DABCYL, and widely used to prepare a variety of FRET-based probes that contain DABCYL.


  • FRET probe precursor
  • Carbonyl-reactive

Molecular Weight: 425,41 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a201ddc088ac9cd622dbacbfb57d498b8eb1ea&vid=8b44470c34dbab7b786246b51db75ab5a99819c7&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR