Probenecid ≥95% auf Zellkulturen getestet

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 4-[(dipropylamino)sulphonyl]benzoic acid

20060
AATB20060EA 120 EUR
AATB20060
Probenecid ≥95% auf Zellkulturen getestet
Probenecid

Probenecid is an inhibitor of organic-anion transporters located in cell membranes.

Formel: C₁₃H₁₉NO₄S
Molecular Weight: 285,4 g/mol
MDL: MFCD00038402
CAS-Nummer: 57-66-9
UN: 3249
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR