Probenecid 25 mM in wässriger Lösung stabilisiert, ReadiUse™
Lieferant: AAT BIOQUEST
Synonyme:
4-[(dipropylamino)sulphonyl]benzoic acid
20062
AATB20062EA
230
EUR
AATB20062
Probenecid 25 mM in wässriger Lösung stabilisiert, ReadiUse™
Probenecid
Each vial is diluted with 90 ml buffer to make 2.5 mM final concentration of probenecid for your assays.
- Convenient to use
AAT Bioquest offer the convenient Readiuse™ water-soluble and heat-stable probenecid in the format of a powder, solution or tablet.
Formel:
C₁₃H₁₉NO₄S Molecular Weight: 285,4 g/mol |
MDL:
MFCD00038402 CAS-Nummer: 57-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...