Annexin V-iFluor® 700 Conjugate
Lieferant: AAT BIOQUEST
20077
AATB20077EA
307
EUR
AATB20077
Annexin V-iFluor® 700 Conjugate
Tests
Zelltests
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
Bitte beachten For research use only.