DABCYL C2 maleimide FRET-Akzeptor-Vorläufer

Lieferant: AAT BIOQUEST
Achtung

2008
AATB2008EA 307 EUR
AATB2008
DABCYL C2 maleimide FRET-Akzeptor-Vorläufer
DABCYL C2 maleimide

DABCYL C2 maleimide is the thiol-reactive form of DABCYL, and widely used to prepare a variety of FRET-based probes that contain DABCYL.


  • FRET probe precursor

Formel: C₂₁H₂₁N₅O₃
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d275632bd1af179f2873ffd156f62f084de39e2&vid=61b360e92ef58147634dcb8c5768666c31de058d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR