Annexin V-mFluor™ Violet 450 Conjugate
Lieferant: AAT BIOQUEST
20080
AATB20080EA
307
EUR
AATB20080
Annexin V-mFluor™ Violet 450 Conjugate
Tests
Zelltests
Annexins are a family of proteins that bind to phospholipid membranes in the presence of calcium.
- High stability
- In-house quality control
- Multiple research applications
- Optimized for flow cytometry
Bitte beachten For research use only.